Learn More
Calsequestrin 2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1C10A1, Proteintech
Código de producto. 16888275 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16888275 150 μL 150 microlitros
16898275 20 μL 20 microlitros
2 options
Código de producto. 16888275
Marca: Proteintech 664191IG150UL
Mouse Monoclonal Antibody
Calsequestrin (CASQ) is a Ca2+-binding protein present primarily in junctional sarcoplasmic reticulum of skeletal and cardiac muscle; the cardiac form (CASQ2) is encoded by a separate gene. The primary role of CASQ2 is buffering of the sarcoplasmic reticulum Ca2+ ions, but another role for CASQ2 has emerged recently: CASQ2 regulates the open probability of ryanodine receptor 2 (RyR2). Mutations in CASQ2 cause stress-induced polymorphic ventricular tachycardia, also referred to as catecholaminergic polymorphic ventricular tachycardia 2 (CPVT2), a disease characterized by bidirectional ventricular tachycardia that may lead to cardiac arrest.
Especificaciones
Antígeno Calsequestrin 2
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Monoclonal
Clon 1C10A1
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica CASQ2
N.º de referencia del gen O09161, O14958, O18934, P51868
Alias de gen Calsequestrin 2, CASQ2, PDIB2
Símbolos de los genes Casq2
Especie del huésped Mouse
Inmunógeno Calsequestrin 2 Fusion Protein Ag13246
Método de purificación Protein A
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 12373, 29209, 397436, 845
Especies diana Human, Mouse, Pig, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2a