missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calpain 3 Polyclonal Antibody
Descripción
Recommended positive controls: HeLa. Predicted reactivity: Mouse (94%), Rat (95%), Xenopus laevis (82%), Pig (94%), Rabbit (89%), Chicken (86%), Sheep (94%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Calpain, an intracellular calcium-dependent protease that cleaves cytoskeletal and submembranous proteins, plays a role in cytoskeletal reorganization and muscle protein degradation. Calpain is a heterodimer composed of a small regulatory subunit and one of three large catalytic subunits, designated calpain 1, calpain 2 and calpain 3. Calpain 3 (calpain p94) is a muscle-preferred calcium activated neutral protease that localizes to the nucleus. Mutations involving the calpain 3 gene are associated with limb-girdle muscle dystrophy type 2A, a form of autosomal recessive and progressive neuromuscular diso4. Calpastatin regulates calpain by inhibiting both the proteolytic activity of calpain and its binding to membranes. Calpastatin exists in two types, tissue type and erythrocyte type, resulting from both alternative splicing and proteolytic processing.
Especificaciones
Especificaciones
| Antígeno | Calpain 3 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.01% thimerosal; pH 7 |
| génica | Capn3 |
| N.º de referencia del gen | P20807 |
| Alias de gen | AI323605; calcium-activated neutral proteinase 3; calpain 3; calpain 3, (p94); calpain L3; Calpain p94; calpain p94, large [catalytic] subunit; calpain, large polypeptide L3; calpain-3; CANP 3; CANP3; CANPL3; Capa3; Capa-3; CAPN 3; CAPN3; LGMD 2A; LGMD2; LGMD2A; Lp82; Lp84; Lp85; MGC10767; MGC11121; MGC14344; MGC4403; Muscle-specific calcium-activated neutral protease 3; muscle-specific calcium-activated neutral protease 3 large subunit; NCL1; nCL-1; New calpain 1; p94; skeletal muscle specific calpain p94 |
| Símbolos de los genes | Capn3 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?