Learn More
Calnexin Antibody (1C2.2D11), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NBP2-36570AF405
Código de producto. 18101989
  • ONLINE EXCLUSIVE PRICE
    457.00€
    431.55€ / 0.10 ml
    Ahorro 25.45€
    5% Descuento
Descripción
Calnexin Monoclonal specifically detects Calnexin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Calnexin
Monoclonal
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
Mouse
Protein G purified
RUO
Primary
Human
Purified
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
1C2.2D11
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular)
P27824
CANX
Partial recombinant human Calnexin protein (between amino acids 1-300). [UniProt P27824]
0.1 mL
ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision
821
Store at 4C in the dark.
IgG2b κ
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones