missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Calcitonin, human, 96%

Product Code. p-5384607
Change view
Click to view available options
Cantidad:
0,5 mg
1 mg
Unit Size:
0.50 miligramo
1 miligramo
Product Code. Cantidad unitSize
15495449 0,5 mg 0.50 miligramo
15405459 1 mg 1 miligramo
2 options
This item is not returnable. View return policy

Product Code. 15495449

Brand: Thermo Scientific Chemicals J63192.!D

Solicitar formato a granel o personalizado

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Para aplicaciones de biología molecular y celular

Decreases blood calcium and phosphate due to inhibition of resorption by osteoblasts and osteocytes. A carrier peptide that can be used to internalize fusion proteins

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Especie Human
Método de purificación HPLC
Pureza ≥97%
Nombre común Human C carcinoma
Contenido y almacenamiento -30°C to -10°C
Formato Polvo
Para utilizar con (aplicación) Decreases blood calcium and phosphate due to inhibition of resorption by osteoblasts and osteocytes. A carrier peptide that can be used to internalize fusion proteins
Peso molecular 3417.90
Tipo de producto Calcitonin, human, ≥97%
Cantidad 0,5 mg
Estado normativo RUO - research use only
Secuencia Calcitonin M
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.