Learn More
CACNB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16841054 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16841054 20 μL 20 microlitros
16831054 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16841054 Proveedor Proteintech N.º de proveedor 130391AP20UL
Rabbit Polyclonal Antibody
The expression of multiple classes of voltage dependent calcium channels (VDCC′s) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCC′s exist before birth which is consistent with a role in migration, most N-Type VDCC′s subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.
Especificaciones
Antígeno CACNB1
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.27 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica CACNB1
N.º de referencia del gen P54283, Q02641, Q8R3Z5
Alias de gen CAB1, CACNB1, CACNLB1, CCHLB1
Símbolos de los genes Cacnb1
Especie del huésped Rabbit
Inmunógeno CACNB1 Fusion Protein Ag3693
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 12295, 50688, 782
Especies diana Rat, Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG