Learn More
CACNB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13039-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The expression of multiple classes of voltage dependent calcium channels (VDCC′s) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCC′s exist before birth which is consistent with a role in migration, most N-Type VDCC′s subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.Especificaciones
| CACNB1 | |
| Polyclonal | |
| Unconjugated | |
| CACNB1 | |
| CAB1, CACNB1, CACNLB1, CCHLB1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12295, 50688, 782 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P54283, Q02641, Q8R3Z5 | |
| Cacnb1 | |
| CACNB1 Fusion Protein Ag3693 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |