missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CA7 Polyclonal Antibody

Description
CA7 Polyclonal Antibody for Western Blot, IHC (P)
Carbonic anhydrases are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. This gene encodes a glycosylphosphatidyl-inositol-anchored membrane isozyme expressed on the luminal surfaces of pulmonary capillaries and proximal renal tubules. Its exact function is not known; however, it may have a role in inherited renal abnormalities of bicarbonate transport.
Specifications
Specifications
| Antígeno | CA7 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1.2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| génica | Ca7 |
| N.º de referencia del gen | P43166 |
| Alias de gen | AV343731; CA7; Car7; Carbonate dehydratase VII; carbonic anhydrase 7; Carbonic anhydrase VII; carbonic dehydratase VII; CAVII; CA-VII |
| Símbolos de los genes | Ca7 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?