Learn More
CA3 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 3C10A2, Proteintech
Código de producto. 16828645 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16828645 20 μL 20 microlitros
16818645 150 μL 150 microlitros
2 options
Código de producto. 16828645
Marca: Proteintech 666081IG20UL
Mouse Monoclonal Antibody
Carbonic anhydrase III (CAIII) is a member of carbonic anhydrase isozymes. These carbonic anhydrases are a class of metalloenzymes that catalyze the reversible hydration of carbon dioxide and are differentially expressed in a number of cell types. The expression of the CA3 gene is strictly tissue specific and present at high levels in skeletal muscle and much lower levels in cardiac and smooth muscle. A proportion of carriers of Duchenne muscle dystrophy have a higher CA3 level than normal.
Especificaciones
Antígeno CA3
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Monoclonal
Clon 3C10A2
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica CA3
N.º de referencia del gen P07451, P14141, P16015, Q5S1S4
Alias de gen CA III, CA3, CAIII, Car3, Carbonate dehydratase III, Carbonic anhydrase 3, Carbonic anhydrase III
Símbolos de los genes Ca3
Especie del huésped Mouse
Inmunógeno CA3 Fusion Protein Ag7513
Método de purificación Protein A
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 12350, 494016, 54232, 761
Especies diana Pig, Human, Rat, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2b