missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C5orf64 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15586482
Change view
Click to view available options
Cantidad:
100 μL
Unit Size:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
15586482 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15586482 Supplier Invitrogen™ Supplier No. PA544396

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: MLAPLFLCCL RNLFRKLISF QPPQLGRTNM HYSKLPRTAI ETEFKQNVGP Sequence homology: Human: 100%.

The specific function of the protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno C5orf64
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 2% sucrose and 0.09% sodium azide
génica C5orf64
N.º de referencia del gen Q2M2E5
Alias de gen C5orf64; chromosome 5 open reading frame 64; uncharacterized protein C5orf64
Símbolos de los genes C5orf64
Especie del huésped Rabbit
Inmunógeno Synthetic peptide directed towards the N-terminal of human FLJ37543 (aa 1-50).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 285668
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.