Learn More
DiscoverX C3a Receptor Agonist
Código de producto. 17362509
Encompass
Click to view available options
:
1 mg
Código de producto. 17362509
Marca: DiscoverX 921187
C3a Receptor Agonist is an agonist for C3AR1. Provided as 1mg of dry powder.
C3a Receptor Agonist is an agonist of the complement component 3a receptor 1 (C3AR1), a G protein-coupled receptor protein that is involved in the complement system. May have a role in chemotaxis, inflammation, and anaphylaxis. Provided as 1mg of dry powder.
Especificaciones
Cantidad 1 mg