Learn More
C20orf3/APMAP Mouse anti-Human, Unconjugated, Clone: 1C6C2, Proteintech
Tienda Proteintech ProductosDescripción
The C20orf3/APMAP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human C20orf3/APMAP. This antibody recognizes human, mouse antigen. The C20orf3/APMAP antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
Especificaciones
| Antígeno | C20orf3/APMAP |
| Aplicaciones | Western Blot, ELISA |
| Clasificación | Monoclonal |
| Clon | 1C6C2 |
| Concentración | 1000 μg/mL |
| Conjugado | Unconjugated |
| Especie del huésped | Mouse |
| Cantidad | 20 μL |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 57136 |
| Mostrar más |