Learn More
C20orf3/APMAP Mouse anti-Human, Unconjugated, Clone: 1C6C2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67464-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The C20orf3/APMAP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human C20orf3/APMAP. This antibody recognizes human, mouse antigen. The C20orf3/APMAP antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
| C20orf3/APMAP | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse |
| Western Blot, ELISA | |
| 1C6C2 | |
| Unconjugated | |
| 150 μL | |
| 57136 | |
| IgG1 |