Learn More
c-Kit/CD117 Rabbit anti-Human, Polyclonal, Proteintech
Código de producto. 16898564 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16898564 20 μL 20 microlitros
16888564 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16898564 Proveedor Proteintech N.º de proveedor 186961AP20UL
Rabbit Polyclonal Antibody
KIT may be involved in testicular germ cell tumor (TGCT). A common maligcy in males representing 95% of all testicular neoplasms. TGCTs have various pathologic subtypes including: unclassified intratubular germ cell neoplasia, seminoma (including cases with syncytiotrophoblastic cells), spermatocytic seminoma, embryonal carcinoma, yolk sac tumor, choriocarcinoma, and teratoma. Note=The gene represented in this entry may be involved in disease pathogenesis.
Especificaciones
Antígeno c-Kit/CD117
Aplicaciones Immunocytochemistry, Immunofluorescence, Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.23 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica KIT
N.º de referencia del gen P10721
Alias de gen C Kit, CD117, KIT, PBT, Proto oncogene c Kit, SCFR, Tyrosine protein kinase Kit
Símbolos de los genes KIT
Especie del huésped Rabbit
Inmunógeno Peptide
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3815
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG