Learn More
Invitrogen™ C/EBP beta Monoclonal Antibody (18F8)

Descripción
MA1-826 detects C/EBP beta protein in human, mouse, and rat samples. MA1-826 only detects the LAP* isoform of C/EBP beta. MA1-826 has successfully been used in Western blot, immunofluorescent and immunoprecipitation procedures. By Western blot, this antibody detects a 37 kDa protein representing LAP* of C/EBP beta (N-terminus fragment) in transfected 293T cells. The MA1-826 immunogen is a synthetic peptide corresponding to residues M(1) H R L L A W D A A C L P P P A A F P K L H(21) of mouse C/EBP beta. This peptide (Cat. # PEP-243) is available for use in neutralization and control experiments.
Especificaciones
Especificaciones
| Antígeno | C/EBP beta |
| Aplicaciones | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 18F8 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1mg/mL BSA and 0.05% sodium azide |
| génica | Cebpb |
| N.º de referencia del gen | P17676, P21272, P28033 |
| Alias de gen | AGP/EBP; C EBP; c/EBP beta; C/EBPb; C/EBPbeta; C/EBP-beta; C/EBP-related protein 2; CCAAT enhancer binding protein beta; CCAAT/enhancer binding protein (C/EBP), beta; CCAAT/enhancer binding protein beta; CCAAT/enhancer-binding protein beta; CEBPB; Crp2; Il6dbp; IL-6DBP; interleukin 6-dependent DNA-binding protein; interleukin-6-dependent-binding protein; LAP; LIP; Liver activator protein; Liver-enriched inhibitory protein; Liver-enriched transcriptional activator; Nfil6; Nf-il6; NF-M; nuclear factor NF-IL6; nuclear factor of interleukin 6; nuclear factor-IL6; nuclear protein Il6; OTTMUSP00000017503; PP9092; Sfb; SF-B; Silencer factor B; TCF5; TCF-5; transcription factor 5; transcription factor C/EBP beta |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.