missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BZLF1 Polyclonal Antibody

Descripción
BZLF1 Polyclonal Antibody for Western Blot, ELISA
Transcription factor that acts as a molecular switch to induce the transition from the latent to the lytic or productive phase of the virus cycle (PubMed:8404860) (Probable). Mediates the switch from the latent to the lytic cycle of infection in cells containing a highly methylated viral genome (PubMed:34893887, PubMed:15361873). Probably binds to silenced chromatin and recruits host chromatin-remodeling enzymes (PubMed:30926617). Regulates this switch by binding to 2 types of ZEBRA response elements (ZREs): the CpG-free AP-1 like elements (latency) and the methylated CpG-containing elements (lytic replication) (PubMed:34893887, PubMed:22022468) (Probable). Activates preferentially the methylated forms of the viral lytic R (BRLF1) and Na (BRRF1) gene promoters, the latters being the first genes activated during Z-mediated reactivation in latently infected cells (PubMed:19325883, PubMed:15361873). BZLF1 and BRLF1 act together to trigger lytic replication (PubMed:10856251, PubMed:15361873). Also binds the lytic origin of replication, oriLyt (PubMed:20808903). Induces G1 cell cycle arrest by stabilizing the host CCAAT/enhancer binding protein CEBPA (PubMed:15078966). This function is important because the lytic cycle preferentially takes place in host cells arrested in G1 (PubMed:15078966).
Especificaciones
Especificaciones
| Antígeno | BZLF1 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 4.9 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 |
| génica | BZLF1 |
| N.º de referencia del gen | P03206 |
| Alias de gen | Trans-activator protein BZLF1 |
| Símbolos de los genes | BZLF1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?