Descripción
The protein encoded by this gene plays a crucial role in B-cell development. Mutations in this gene cause X-linked agammaglobulinemia type 1, which is an immunodeficiency characterized by the failure to produce mature B lymphocytes, and associated with a failure of Ig heavy chain rearrangement.
Especificaciones
Especificaciones
| Antígeno | BTK |
| Aplicaciones | Immunoprecipitation, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.13 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | BTK |
| N.º de referencia del gen | P35991, Q06187 |
| Alias de gen | AGMX1, AT, ATK, B cell progenitor kinase, BPK, Bruton tyrosine kinase, BTK, IMD1, PSCTK1, Tyrosine protein kinase BTK, XLA |
| Símbolos de los genes | Btk |
| Mostrar más |