missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
This antibody is specific to PAX5. It will not cross react with other PAX members.
PAX5 is a member of the paired box (PAX) family of transcription factors. The PAX proteins are important regulators in early development and alterations in the expression of their genes are thought to contribute to neoplastic transformation. PAX5 is the B-cell lineage specific activator protein (BSAP) that is expressed at early, but not late stages, of B-cell differentiation. Its expression has also been detected in developing CNS and testis, therefore, PAX5 may not only play an important role in B-cell differentiation, but also in neural development and spermatogenesis. Mutations in the gene can result in leukemia, acute lymphoblastic 3.
Especificaciones
Especificaciones
| Antígeno | BSAP/PAX5 |
| Aplicaciones | Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.53 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | PAX5 |
| N.º de referencia del gen | Q02548 |
| Alias de gen | BSAP, BSAP,PAX5, paired box 5, Paired box protein Pax 5, PAX5 |
| Símbolos de los genes | PAX5 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?