missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human BRF2. It recognizes a 50-60 kDa band in WB analysis.
This gene encodes one of the multiple subunits of the RNA polymerase III transcription factor complex required for transcription of genes with promoter elements upstream of the initiation site. The product of this gene, a TFIIB-like factor, is directly recruited to the TATA-box of polymerase III small nuclear RNA gene promoters through its interaction with the TATA-binding protein.
Especificaciones
Especificaciones
| Antígeno | BRF2 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.29 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | BRF2 |
| N.º de referencia del gen | Q3UAW9, Q4V8D6, Q9HAW0 |
| Alias de gen | B related factor 2, BRF 2, BRF2, BRFU, hBRFU, hTFIIIB50, TFIIIB50 |
| Símbolos de los genes | Brf2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?