Learn More
Invitrogen™ Brevican Polyclonal Antibody
GREENER_CHOICE
Código de producto. 13209328
Change view
Click to view available options
Cantidad:
400 μL
Tamaño de la unidad:
100 microgramos
13209328 400 μL 100 microgramos
1 options
Código de producto. 13209328
Marca: Invitrogen™ PA525808
Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine based on sequence homology.
Brevican is the most abundant chondroitin sulfate proteoglycan in the extracellular matrix of the adult brain. It is a member of the lectican family of aggregating extracellular matrix proteoglycans that bear chondroitin sulfate (CS) chains. It is highly expressed in the central nervous system and is thought to stabilize synapses and inhibit neural plasticity. Brevican is secreted from astrocytes and neurons as a 145 kD core protein that bears up to three, covalently-linked, CS chains. It is also is secreted as a 145 kD core protein without CS chains. When cleaved by extracellular glutamyl endopeptidases, the ADAMTSs, a 55 kD N-terminal fragment is formed that contains the unique C terminal epitope EAMESE.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Brevican
Aplicaciones Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.34 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.09% sodium azide; pH 7.4
génica Bcan
N.º de referencia del gen Q96GW7
Alias de gen ALPBRE; Bcan; Behab; Brain-enriched hyaluronan-binding protein; brevican; brevican (brain specific proteoglycan in the aggrecan family); brevican core protein; Chondroitin sulfate proteoglycan 7; chondroitin sulfate proteoglycan BEHAB; CSPG7; UNQ2525/PRO6018
Símbolos de los genes Bcan
Especie del huésped Rabbit
Inmunógeno KLH conjugated synthetic peptide between 715-741 amino acids from the C-terminal region of human BCAN.
Método de purificación Antigen Affinity Chromatography, Protein A
Cantidad 400 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 63827
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG