Descripción
The BRD4 antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human BRD4. This antibody recognizes human antigen. The BRD4 antibody has been validated for the following applications: IP, WB, ELISA analysis.
Especificaciones
Especificaciones
| Antígeno | BRD4 |
| Aplicaciones | Western Blot, Immunoprecipitation |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Especie del huésped | Rabbit |
| Cantidad | 20 μL |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 23476 |
| Especies diana | Human |
| Isotype | IgG |