Learn More
Brady™ BradyPrinter i7100 with Peel Function
Código de producto. 30242845
Click to view available options
Resolución:
300 ppp
600 ppp
Alternate Part Number:
i7100-300-EU-BWS
i7100-600P-EU-BWS
Tamaño de la unidad:
1 set
Código de producto. 30242845
Marca: Brady™ 322880
Solo quedan null
Añadir a la cesta
Añadir a la cesta
The BradyPrinter i7100 industrial thermal transfer benchtop printer is ideal for reliable high volume and accurate printing on sleeves, tags, labels, tapes, cable markers and more.
  • Print on the widest range of materials with confidence, including difficult to print materials
  • Peel configuration automatically peels labels from the liner
  • Prints ultra-sharp on labels as small as 4 mm with high accuracy
  • Designed to print thousands of labels per day, everyday
  • Easy-to-use touch screen interface to adjust printer settings quickly and easily
  • Ships with a licence for the Brady Workstation suite of your preference: Product and Wire Identification, Safety and Facility Identification, or Lab Identification, enabling easy creation of a wide range of labels and signs.
Especificaciones
Tipo de producto Printer Kit
Color Blue, Gray
Tipo de pantalla Color LCD
Material Plastic
Print Technology Transferencia térmica directa
Longitud (imperial) 18.1 in.
Anchura (imperial) 9,9 in
Longitud (métrico) 459.74 mm
Anchura (métrico) 251.46 mm
Altura (imperial) 11,4 in
Altura (métrico) 289.56 mm
Dimensiones (L x An) 459.74 x 251.46 x 289.56 mm
Capacidad de color Monocolour
Tamaño de la pantalla 50.80 x 91.44 mm
Incluye Brady Workstation Suite, Peel Function
Idiomas English, Spanish, German, French, Dutch
Memoria 256 MB RAM, 50 MB IFFS Flash, SD card up to 512 GB
Modelo i7100-600P-EU-BWS
Velocidad de impresión 150.00 mm/sec.
Resolución 600 ppp
Tipo de software Brady Workstation, Windows
Tipos de conector Ethernet, I/O connection, Bluetooth (optional), Wi-Fi (optional), Serial, USB
Sistema operativo Windows™ 7, Windows™ 8, Windows™ 10, Windows™ Server 2012, Windows™ Server 2012 R2, Windows™ Server 2016
Peso (imperial) 22.05 lb.
Peso (métrico) 10 kg
Certificaciones/conformidad CE, FCC class A, FCC class CB, CCC, UL
Garantía 2 years
Detalles de la fuente de alimentación AC Adaptor
Tensión 100/240 V
Keyboard Type None
Alternate Part Number i7100-600P-EU-BWS
Tipo de tapón EU