Learn More
Hecht Karl™ Borosilicate Glass 3.3 Automatic Pipets
missing translation for 'orderingAttributeHoverText'
Volumen (métrico):
10 mL
25 mL
50 mL
missing translation for 'unitSize'
cada uno
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Incluye | Standard Ground NS 29/32 |
| Cantidad | 1/pack |
| Volumen (métrico) | 25 mL |
| Tipo | Automatic Pipette |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction