Learn More
Boekel Scientific™ Replicators
GREENER_CHOICE
Código de producto. 10464225
Change view
Click to view available options
:
96 pin replicator
Código de producto.
10464225 96 pin replicator
1 options
Código de producto. 10464225 Proveedor Boekel Scientific™ N.º de proveedor 140500
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Save time in transferring small volumes from one 96- or 48-well microplate to another
  • For replicating cosmid and VAC libraries, among other uses
  • To use, dip durable stainless-steel, precision-aligned pins into standard 96- or 48-well plate; lift and move replicator to a second microplate
  • Replicator frame is anodized aluminum with comfortable nylon handle
  • Autoclavable or flame sterilizable
  • With anodized aluminum storage pan that can hold cleansing pads to remove build-up on pins or ethanol for flame sterilization
Información de órdenes
Dimensions: 3L x 4.5W x 4 in.H (7.6 x 11.4 x 10cm); pins measure 1/16L x 1/16 in.W (1.5 x 1.5mm).
Especificaciones
Material Stainless Steel
Esterilidad Sterile
Tipo de producto 96 Pin Replicator
Esterilizables en autoclave Yes
Altura (métrico) 11 cm
Anchura (imperial) 4.5 in.
Anchura (métrico) 11 cm
Altura (imperial) 4.5 in.
Para utilizar con (aplicación) Colony hybridization, antibiotic sensitivity testing, phage typing, transfer small volumes of inoculums from microplates to daughter plates or membranes, replication of YACs and cosmid libraries