missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOB.1 (B-Cell Marker) Mouse, Clone: BOB1/2424, enQuireBio™
Specifications
Specifications
| Antígeno | BOB.1 (B-Cell Marker) |
| Aplicaciones | Immunohistochemistry |
| Localización celular | Nuclear |
| Clasificación | Monoclonal |
| Clon | BOB1/2424 |
| Concentración | 200μg/mL |
| Conjugado | Unconjugated |
| Formulación | 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Recombinant human BOB1 (POU2AF1) protein fragment (apprx. aa 148-255) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?