missing translation for 'onlineSavingsMsg'
Learn More

BMS-986020, MedChemExpress

Código de producto. 16465837
Encompass_Preferred
Change view
Click to view available options
Cantidad:
1 mg
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
1 miligramo
10 miligramos
100 miligramos
5 miligramos
50 miligramos
Código de producto. Cantidad unitSize
16465837 5 mg 5 miligramos
16499656 50 mg 50 miligramos
16494396 100 mg 100 miligramos
16440679 1 mg 1 miligramo
16433108 10 mg 10 miligramos
5 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16465837

Marca: Medchem Express HY100619

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress BMS-986020 (AM152) is a high-affinity and selective lysophosphatidic acid receptor 1 (LPA1) antagonist. BMS-986020 inhibits bile acid and phospholipid transporters with IC50s of 4.8 μM, 6.2 μM, and 7.5 μM for BSEP, MRP4, and MDR3, respectively. BMS-986020 has the potential for the treatment of idiopathic pulmonary fibrosis (IPF).

BMS-986020 (AM152) is a high-affinity and selective lysophosphatidic acid receptor 1 (LPA1) antagonist. BMS-986020 inhibits bile acid and phospholipid transporters with IC50s of 4.8 μM, 6.2 μM, and 7.5 μM for BSEP, MRP4, and MDR3, respectively. BMS-986020 has the potential for the treatment of idiopathic pulmonary fibrosis (IPF).

Specifications

Nombre del producto químico o material BMS-986020
Forma física Solid
CAS 1257213-50-5
Color Blanco
Cantidad 5 mg
Notas de grado de pureza Research
Fórmula molecular C29H26N2O5
Para utilizar con (aplicación) COVID-19-immunoregulation
Sinónimo AM152
Información de solubilidad DMSO : 250 mg/mL (518.10 mM; Need ultrasonic)
SMILES O=C(C1(C2=CC=C(C3=CC=C(C4=C(NC(O[C@@H](C5=CC=CC=C5)C)=O)C(C)=NO4)C=C3)C=C2)CC1)O
Peso molecular (g/mol) 482.53
Formula Weight (peso de la fórmula) 482.53
Porcentaje de pureza 98.0%
Grado Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.