missing translation for 'onlineSavingsMsg'
Learn More

BMS-345541, MedChemExpress

Código de producto. 16426737
Encompass_Preferred
Click to view available options
Cantidad:
5 mg
50 mg
Tamaño de la unidad:
5 miligramos
50 miligramos
Este artículo no se puede devolver. View return policy

Product Code. 16426737

missing translation for 'mfr': Medchem Express HY10519

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress BMS-345541 is a selective inhibitor of the catalytic subunits of IKK (IKK-2 IC50=0.3 μM, IKK-1 IC50=4 μM). BMS-345541 binds at an allosteric site of IKK.

BMS-345541 is a selective inhibitor of the catalytic subunits of IKK (IKK-2 IC50=0.3 μM, IKK-1 IC50=4 μM). BMS-345541 binds at an allosteric site of IKK.

Specifications

Nombre del producto químico o material BMS-345541
Forma física Solid
CAS 445430-58-0
Color Blanco
Cantidad 5 mg
Notas de grado de pureza Research
Fórmula molecular C14H17N5
Para utilizar con (aplicación) Cancer-Kinase/protease
Información de solubilidad DMSO : 10 mg/mL (39.17 mM; Need ultrasonic)
SMILES CC1=CN=C2C(NCCN)=NC3=CC=C(C)C=C3N21
Peso molecular (g/mol) 255.32
Formula Weight (peso de la fórmula) 255.32
Porcentaje de pureza 99.57%
Grado Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.