missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Blood Group M Antigen Monoclonal Antibody (GH-9)

Descripción
MA1-7676 detects M human blood protein from human samples. MA1-7676 has been successfully used in agglutination applications. The MA1-7676 immunogen is a M human blood protein.
The MNS antigen system is a human blood group system based upon two genes (glycophorin A and glycophorin B) on chromosome 4. The MN blood group system is under the control of an autosomal locus found on chromosome 4, with two alleles designated L-M and L-N. The blood type is due to a glycoprotein present on the surface of red blood cells, which behaves as a native antigen. Phenotypic expression at this locus is codominant because an individual may exhibit either one or both antigenic substances. These antigens were an early discovery and are some of the oldest blood antigens known after the ABO system. Anti-M and anti-N antibodies are usually IgM and are rarely associated with transfusion reactions.
Especificaciones
Especificaciones
| Antígeno | Blood Group M Antigen |
| Aplicaciones | Agglutination |
| Clasificación | Monoclonal |
| Clon | GH-9 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.2% gelatin, 0.5% saccharose and 0.01% sodium azide; pH 7.2 |
| Alias de gen | Blood Group LM; MN Blood Group |
| Especie del huésped | Mouse |
| Inmunógeno | M human blood protein. |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?