Learn More
BLNK Monoclonal Antibody (OTI2B2), TrueMAB™, OriGene
Código de producto. 17579853
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Tamaño de la unidad:
100 microlitros
30 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17579853 30 μL 30 microlitros
17559853 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17579853 Proveedor Origene Technologies N.º de proveedor TA808522S
Mouse Monoclonal Antibody
BLNK Monoclonal antibody specifically detects BLNK in Human samples. It is validated for Western Blot
Especificaciones
Antígeno BLNK
Aplicaciones Western Blot
Clasificación Monoclonal
Clon OTI2B2
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica AMTN
N.º de referencia del gen Q6UX39
Alias de gen AGM4; B cell adaptor containing SH2 domain; B cell linker; B cell linker protein; BASH; bca; B-cell activation; B-cell adapter containing a SH2 domain protein; B-cell adapter containing a Src homology 2 domain protein; B-cell linker; B-cell linker protein; Blnk; BLNK-S; Cytoplasmic adapter protein; LY57; Ly-57; lymphocyte antigen 57; Lyw-57; MGC111051; SLP65; Slp-65; Src homology [SH2] domain-containing leukocyte protein of 65 kD; Src homology 2 domain-containing leukocyte protein of 65 kDa
Especie del huésped Mouse
Inmunógeno Full length human recombinant protein of human AMTN produced in E.coli.
Método de purificación Affinity chromatography
Cantidad 30 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 401138
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Formulario Liquid
Isotype IgG1