missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Blinatumomab Recombinant Mouse Monoclonal Antibody

Mouse Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA541729
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Blinatumomab is a BiTE-class (bi-specific T-cell engagers) constructed monoclonal antibody indicated for the treatment of Philadelphia chromosome-negative relapsed or refractory B-cell precursor acute lymphoblastic leukemia (ALL). Blinatumomab is manufactured by Amgen Inc. and marketed under the brand Blincyto™ Blinatumomab was approved in December 2014 under the FDA's accelerated approval program, which allows approval of a pharmaceutical to treat a serious or life-threatening disease based on clinical data showing the pharmaceutical has an effect on a surrogate endpoint reasonably likely to predict clinical benefit to patients.
Especificaciones
| Blinatumomab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| AMG103; BITE MT-103; bscCD19xCD3; MEDI-538; MT103 | |
| Human CD19/CD3E. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA | |
| 1.61 mg/mL | |
| 30mM histidine with 10% sucrose, 0.02% Tween 80 and no preservative; pH 5.8 | |
| Mouse | |
| Metal-chelate chromatography | |
| RUO | |
| Human | |
| Antibody | |
| scFv κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido