missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BK Virus VP1 Monoclonal Antibody (4942)
Descripción
BK Virus VP1 Monoclonal Antibody for ELISA, ICC/IF
BK Virus VP1 forms an icosahedral capsid with a T=7 symmetry and a 50 nm diameter. The capsid is composed of 72 pentamers linked to each other by disulfide bonds and associated with VP2 or VP3 proteins. Interacts with gangliosides GT1b and GD1b containing terminal alpha2-8-linked sialic acids on the cell surface to provide virion attachment to target cell. This attachment induces virion internalization predominantly through caveolin-mediated endocytosis and traffics to the endoplasmic reticulum. Inside the endoplasmic reticulum, the protein folding machinery isomerizes VP1 interpentamer disulfide bonds, thereby triggering initial uncoating. Next, the virion uses the endoplasmic reticulum-associated degradation machinery to probably translocate in the cytosol before reaching the nucleus. Nuclear entry of the viral DNA involves the selective exposure and importin recognition of VP2/Vp3 nuclear localization signal. In late phase of infection, neo-synthesized VP1 encapsulates replicated genomic DNA in the nucleus, and participates in rearranging nucleosomes around the viral DNA.
Especificaciones
Especificaciones
| Antígeno | BK Virus VP1 |
| Aplicaciones | ELISA, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 4942 |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.1% sodium azide; pH 7.2 |
| Alias de gen | BKV; BKV VP1 |
| Especie del huésped | Mouse |
| Método de purificación | Protein A |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?