missing translation for 'onlineSavingsMsg'
Learn More

Birch pollen Bet V1 Antibody (BV16), CoraFluor™ 1, Novus Biologicals™

Código de producto. 30143247
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30143247 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30143247 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP281093CL1

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Birch pollen Bet V1 Monoclonal antibody specifically detects Birch pollen Bet V1 in Plant samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Birch pollen Bet V1
Aplicaciones ELISA, Western Blot
Clasificación Monoclonal
Clon BV16
Conjugado CoraFluor 1
Formulación PBS
Alias de gen Allergen Bet v I-A, Allergen=Bet v 1-A, BETVI, BETVIA, Major pollen allergen Bet v 1-A
Especie del huésped Rabbit
Inmunógeno Birch pollen Bet V1-A, a major allergen of the European white birch.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
Especies diana Plant
Contenido y almacenamiento Store at 4°C in the dark. Do not freeze.
Formulario Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.