Learn More
BIRC5/Survivin Monoclonal Antibody (OTI2D3), TrueMAB™, OriGene
Descripción
BIRC5/Survivin Monoclonal antibody specifically detects BIRC5/Survivin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Especificaciones
Especificaciones
| Antígeno | BIRC5/Survivin |
| Aplicaciones | Flow Cytometry, Immunocytochemistry, Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI2D3 |
| Concentración | 0.54 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | BCL10 |
| N.º de referencia del gen | O95999 |
| Alias de gen | AAC-11; AP14; Api4; apoptosis inhibitor 4; Apoptosis inhibitor survivin; baculoviral IAP repeat containing 5; baculoviral IAP repeat-containing 5; baculoviral IAP repeat-containing 5 (survivin); baculoviral IAP repeat-containing protein 5; BIRC 5; BIRC5; EPR 1; EPR-1; Iap4; inhibitor of apoptosis; survivin; survivin variant 3 alpha; survivin40; SVV; TIAP |
| Mostrar más |