missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Éster de N-hidroxisucinimida de (+)-biotina, 98 %

Product Code. p-4228375 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Cantidad:
250 mg
1 g
Envase:
Frasco de vidrio
Unit Size:
1 g
250 miligramos
Product Code. Cantidad Envase unitSize
11986841 250 mg Frasco de vidrio 250 miligramos
11976841 1 g Frasco de vidrio 1 g
2 options
This item is not returnable. View return policy

Product Code. 11986841

Brand: Thermo Scientific Chemicals 439302500

Solicitar formato a granel o personalizado

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

CAS: 35013-72-0 | C14H19N3O5S | 341.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35013-72-0
Fórmula molecular C14H19N3O5S
Peso molecular (g/mol) 341.39
Clave InChI YMXHPSHLTSZXKH-RVBZMBCESA-N
Sinónimo +-biotin n-hydroxysuccinimide ester, biotin-nhs, biotin nhs, 2,5-dioxopyrrolidin-1-yl 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanoate, n-hydroxysuccinimidobiotin, d-biotinyl-osu, biotin-osu, n-succinimidyl d-biotinate, bhse, biotin n-hydroxysuccinimide ester
PubChem CID 6710714
Nombre IUPAC (2,5-dioxopirrolidin-1-il) 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahidrotieno[3,4-d]imidazol-4-il]pentanoato
SMILES C1CC(=O)N(C1=O)OC(=O)CCCCC2C3C(CS2)NC(=O)N3

Specifications

Nombre del producto químico o material (+)-Biotin N-hydroxysuccinimide ester
% mín. CAS 97.5
% máx. CAS 100.0
Punto de fusión 202.3°C to 202.7°C
Color Blanco
Análisis elemental 48.03 to 50.49% C, 12.00 to 12.62% N
Espectro infrarrojo Auténtico
Intervalo de porcentaje de ensayo 98%
Envase Frasco de vidrio
Cantidad 250 mg
Formula Weight (peso de la fórmula) 341.39
Porcentaje de pureza 98%
Forma física Polvo
Show More Show Less
Product Identifier
  • (+)-Biotin N-hydroxysuccinimide ester, 98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.