Learn More
Ansell BioClean™ Sterile Nitrile RABS/Isolator Sleeve
Descripción
- Assured quality: The BioClean™ Sterile Nitrile RABS/Isolator Sleeve is triple bagged, for assured integrity, and 100% visually inspected, ensuring high levels of quality
- Enhanced versatility: These nitrile protective sleeves are compatible with a wide range of cuff systems, extending protective coverage to the arm
- Specialized protection: In addition, they are autoclave compatible, offering wearers' arm protection in highly pressurized, high-temperature conditions
- Added benefits: These arm protection sleeves also feature extremely low levels of particles and offer excellent ESD properties
Especificaciones
Especificaciones
| Certificaciones/conformidad (Europa) | EC 1935/2004, ISO 11137-1:2006, EN 420:2003 + A1:2009 |
| Chlorinated | No |
| Cleanroom Class | Class 10 (ISO 4) |
| Color | White |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Nitrile |
| Tipo de producto | Isolator, RABS Sleeve |
| Tamaño | 10 |
| Grosor (imperial) | 20 mil |
| Mostrar más |