Learn More
Ansell BIOCLEAN™ LEGEND BLHN Non-Sterile Disposable Latex Cleanroom Glove
Código de producto. 30040342
Change view
Click to view available options
Tamaño:
10
6.5
7
7,5
8
8,5
9
Tamaño de la unidad:
200 pares
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Tamaño unitSize
30040342 8,5 200 pares
30040343 6.5 200 pares
30040330 7 200 pares
30040344 7,5 200 pares
30040338 8 200 pares
30040331 9 200 pares
30040339 10 200 pares
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Código de producto. 30040342 Proveedor Ansell N.º de proveedor BLHN85
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Hand protection with superb comfort and flexibility, textured finger surface with a beaded cuff for cleanroom gloves with grip, and double-donning-friendly design for enhanced convenience and protection.
  • Elevated comfort: BioClean™ Legend BLHN cleanroom gloves' flexible latex material and anatomical design ensure everyday comfort
  • Enhanced features: These non-sterile latex gloves feature a textured finger surface, for enhanced grip performance, while their beaded cuff design provides strength and stability
  • Added protection: BioClean™ Legend BLHN cleanroom gloves are also designed to make double-donning easy for wearers, granting them an extra layer of protection
  • Reduced contamination risks: Their non-particulating EasyTear packaging further minimizes the risk of contaminants entering the cleanroom environment
TRUSTED_SUSTAINABILITY
Especificaciones
Certificaciones/conformidad (Europa) CE 0493, EN 421:2010, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, EN 455 Part 2, Food Contact, Category III, UKCA
Chlorinated Sí
Cleanroom Class Class 10 (ISO 4)
Accelerator Free No
AQL 1.5
Color Natural
Desechables Single-use
Acabado exterior Punta de los dedos texturizada
Finger Thickness (English) 9.44 mil
Finger Thickness (Metric) 0.24 mm
Hand Específico
Latex Free No
Material Látex
Palm Thickness (English) 7.87 mil
Palm Thickness (Metric) 0.2 mm
En polvo sin polvo
Tipo de producto Cleanroom Gloves
Tamaño 8,5
Esterilidad No estéril
Grosor (imperial) 7.87 mil
Grosor (métrico) 0.2 mm
Longitud (imperial) 11.61 in.
Longitud (métrico) 295 mm