Learn More
Invitrogen™ Bimagrumab Recombinant Monoclonal Antibody
Código de producto. 30283111
Change view
Click to view available options
Cantidad:
1 mg
100 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
30283111 1 mg 1 miligramo
30284065 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 30283111 Proveedor Invitrogen™ N.º de proveedor MA558424
Recombinant Monoclonal Antibody
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Bimagrumab has been used in trials studying the treatment and supportive care of Sarcopenia, Skeletal Muscle, Mechanical Ventilation, Sporadic Inclusion Body Myositis, and Sporadic Inclusion Body Myositis (sIBM), among others.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Bimagrumab
Aplicaciones ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Clasificación Recombinant Monoclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Alias de gen BYM338
Especie del huésped Human
Método de purificación Protein A
Cantidad 1 mg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG1