Learn More
BHMT Mouse anti-Human, Clone: OTI10B3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies CF500960
Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a cytosolic enzyme that catalyzes the conversion of betaine and homocysteine to dimethylglycine and methionine, respectively. Defects in this gene could lead to hyperhomocystinemia, but such a defect has not yet been observed.Especificaciones
| BHMT | |
| Monoclonal | |
| Unconjugated | |
| BHMT | |
| BHMT1, HEL-S-61p | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 635 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI10B3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q93088 | |
| BHMT | |
| Full length human recombit protein of human BHMT produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.