Learn More
BHLHE40 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16887894 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16887894 150 μL 150 microlitros
16897894 20 μL 20 microlitros
2 options
Código de producto. 16887894
Marca: Proteintech 178951AP150UL
Rabbit Polyclonal Antibody
This gene encodes a member of the nucleoporin family. The protein is localized to the nuclear rim and is an essential component of the nuclear pore complex (NPC). All molecules entering or leaving the nucleus either diffuse through or are actively transported by the NPC. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
Especificaciones
Antígeno BHLHE40
Aplicaciones Immunoprecipitation, Western Blot
Clasificación Polyclonal
Concentración 0.35 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica BHLHE40
N.º de referencia del gen O14503, O35185, O35780
Alias de gen BHLHB2, BHLHE40, DEC1, SHARP 2, SHARP2, STRA13, Stra14
Símbolos de los genes BHLHE40
Especie del huésped Rabbit
Inmunógeno BHLHE40 Fusion Protein Ag12236
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 20893, 79431, 8553
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG