Learn More
Tocris Bioscience™ BGC 20-1531 hydrochloride
High affinity and selective EP4 antagonist
Marca: Tocris Bioscience™ 5327/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
BGC 20-1531 hydrochloride is a high affinity and selective EP4 antagonist (Ki = 3 nM). Exhibits >2500-fold selectivity for EP4 over EP2 and EP3. Also selective over a range of other prostanoid receptors, ion channels, transporters and enzymes. Inhibits PGE2-induced vasodilation of middle cerebral and meningeal arteries in vitro, and carotid blood flow in vivo. Orally bioavailable.Especificaciones
| 4-[[4-(5-Methoxy-2-pyridinyl)phenoxy]methyl]-5-methyl-N-[(2-methylphenyl)sulfonyl]-2-furancarboxamide hydrochloride | |
| PGN 1531 | |
| 10 mg |