missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ beta Synuclein Monoclonal Antibody (6A10)
Descripción
beta Synuclein Monoclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
Synuclein family of proteins is abundantly expressed in neuronal cytosol and presynaptic terminals. In vertebrates they are encoded by three different genes. Synucleins have been specifically implemented in three major diseases: Alzheimer's (AD), Parkinson's PD) and breast cancer. In AD, a peptide derived from alpha synuclein forms an intrinsic component of plaque amyloid. In PD, an alpha synuclein accumulates in Lewy bodies. An allele of alpha synuclein has been linked to many familial cases of PD. In breast cancer increased expression of gamma synuclein correlates with the disease progression. Synucleins appear to be involved in the membrane plasticity in developing song control system of songbirds.
Specifications
Specifications
| Antígeno | beta Synuclein |
| Aplicaciones | Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 6A10 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 5mM sodium azide |
| génica | SNCB |
| N.º de referencia del gen | P33567, Q16143, Q63754, Q91ZZ3 |
| Alias de gen | AI838531; betaSYN; beta-synuclein; Phosphoneuroprotein 14; PNP 14; Sncb; synuclein beta; synuclein, beta |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?