missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Arrestin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
401.00€ - 672.00€
Especificaciones
| Antígeno | beta-Arrestin 2 |
|---|---|
| Dilución | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Aplicaciones | Immunofluorescence |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
|
18675664
|
Novus Biologicals
NBP3-21381-25ul |
25 μg |
401.00€
25 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
|
18632364
|
Novus Biologicals
NBP3-21381-100ul |
100 μg |
672.00€
100 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Description
beta-Arrestin 2 Polyclonal antibody specifically detects beta-Arrestin 2 in Human samples. It is validated for ImmunofluorescenceSpecifications
| beta-Arrestin 2 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ARB2, ARR2, arrestin 3, Arrestin beta-2, arrestin, beta 2, BARR2, beta-arrestin-2, DKFZp686L0365 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: FVLMHPKPHDHIPLPRPQSAPTPTPPLP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.2, 40% glycerol | |
| 409 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title