Learn More
beta-Actin Mouse anti-Human, Mouse, Rat, Clone: 937215, R&D Systems™

Mouse Monoclonal Antibody has been used in 21 publication
Marca: R&D Systems MAB8929-SP
Descripción
beta-Actin Monoclonal specifically detects beta-Actin in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Especificaciones
| beta-Actin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, betaActin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 60 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| 937215 | |
| Western Blot 0.01 ug/mL, Simple Western 1 ug/mL, Immunocytochemistry 0.1-25 ug/mL | |
| P60709 | |
| ACTB | |
| Peptide containing a sequence at the N-terminus of human beta-Actin Accession # P60709 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat beta-Actin in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.