Learn More
beta-Actin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 13 publications
Marca: Novus Biologicals NB600-505
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
beta-Actin Polyclonal specifically detects beta-Actin in Human, Mouse, Rat, Amphibian samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Electron Microscopy.
Especificaciones
| beta-Actin | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1 | |
| Rabbit | |
| 42 kDa | |
| 0.1 mg | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers | |
| 60 | |
| Human, Mouse, Rat, Amphibian, Bovine, Guinea Pig, Goat, Hamster, Sheep | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay | |
| 1 mg/ml | |
| Western Blot 1:1000-1:4000, ELISA 1:10000-1:40000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:500-1:2000, Immunohistochemistry-Paraffin 1:50 - 1:200, Electron Microscopy | |
| P60709 | |
| ACTB | |
| beta-Actin Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to C-Terminal region near amino acids 350-375 of Human beta Actin. (Uniprot: P60709) | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Expected to cross-react with a wide range of species including most vertebrates and some yeast due to sequence identity. | |
| Store at -20°C. Avoid freeze-thaw cycles. |