Learn More
Invitrogen™ beta-3 Defensin Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA14216
Descripción
PA1-4216 detects the beta-Defensin-3 protein from human samples. PA1-4216 has been successfully used in Western blot and immunohistochemistry procedures. The PA1-4216 immunogen is a synthetic peptide corresponding to residues G(23) I I N T L Q K Y Y C(33) of the 5 kDa human beta Defensin-3 protein.
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].
Especificaciones
| beta-3 Defensin | |
| Polyclonal | |
| Unconjugated | |
| DEFB103A | |
| BD3; BD-3; beta defensin 3; beta defensin-3; beta-defensin 103; Beta-defensin 3; DEFB103; DEFB103A; DEFB103B; DEFB3; DEFB-3; defensin beta 103A; defensin, beta 103; defensin, beta 103A; defensin, beta 3; defensin-like protein; HBD3; hBD-3; HBP3; HBP-3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 414325 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P81534 | |
| DEFB103A | |
| Synthetic peptide corresponding to residues G(23) I I N T L Q K Y Y C(33) of human beta Defensin-3. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.