Learn More
Invitrogen™ Bepranemab Recombinant Monoclonal Antibody
GREENER_CHOICE
Código de producto. 30282776
Change view
Click to view available options
Cantidad:
1 mg
100 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
30282776 100 μg 100 microgramos
30284620 1 mg 1 miligramo
2 options
Código de producto. 30282776
Marca: Invitrogen™ MA559799
Recombinant Monoclonal Antibody
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Bepranemab is an anti-tau monoclonal antibody that targets and binds to tau proteins, facilitating their clearance and reducing tau pathology in neurodegenerative diseases.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Bepranemab Humanized
Aplicaciones ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Clasificación Recombinant Monoclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Alias de gen RG-6416; UCB-0107
Especie del huésped Human
Método de purificación Protein A
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG4SP