Learn More
BEND3 Rabbit anti-Human, Polyclonal, Proteintech
Código de producto. 16842195 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16842195 20 μL 20 microlitros
16832195 150 μL 150 microlitros
2 options
Código de producto. 16842195
Marca: Proteintech 231011AP20UL
Rabbit Polyclonal Antibody
Product No. 23101-1-AP specifically recognizes 85 and 94 kDa isoforms.
Transcriptional repressor which associates with the NoRC (nucleolar remodeling complex) complex and plays a key role in repressing rDNA transcription. The sumoylated form modulates the stability of the NoRC complex component BAZ2A/TIP5 by controlling its USP21-mediated deubiquitination. Binds to unmethylated major satellite DNA and is involved in the recruitment of the Polycomb repressive complex 2 (PRC2) to major satellites ().
Especificaciones
Antígeno BEND3
Aplicaciones Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot
Clasificación Polyclonal
Concentración 0.3 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica BEND3
N.º de referencia del gen Q5T5X7
Alias de gen BEN domain containing 3, BEND3, KIAA1553, RP11 59I9.2
Símbolos de los genes BEND3
Especie del huésped Rabbit
Inmunógeno BEND3 Fusion Protein Ag19137
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 57673
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG