Learn More
Chemsplash Beard Mask Non Woven Disposable
Descripción
Non-Woven Beard Mask
- Material: Non-Woven
- Features:
- Elasticated
- 100% Latex Free
- 18 in.
- PFAS Free
The Disposable Beard Mask is made from a non-woven material, designed as a one-size-fits-all solution. An elasticated strap ensures a secure fit to the face, making it suitable for use in BRC-approved work areas and the pharmaceutical industry.
Especificaciones
Especificaciones
| Tipo | General Purpose Beard Cover |
| Tamaño | One Size |
| Material | Polipropileno |
| Color | Red |
| Desechables | Single-use |
| Sin látex | Sí |
| Longitud (imperial) | 18 in. |
| Longitud (métrico) | 45 cm |