Learn More
BCR Mouse anti-Human, Unconjugated, Clone: 2C2E3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67260-1-IG-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The BCR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BCR. This antibody recognizes human, rat antigen. The BCR antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
| BCR | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat |
| Western Blot, ELISA | |
| 2C2E3 | |
| Unconjugated | |
| 20 μL | |
| 613 | |
| IgG2b |