missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl2 Mouse anti-Mouse, Clone: 1B5B9, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66799-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
BCL-2 is a key regulator of apoptosis that functions to either inhibit or promote cell death. The BCL-2 family members are also characterized by dimerizing to further modulate apoptosis. Bag1, for example, has been found to form a heterodimer with BCL-2 resulting in the enhancement of the anti-apoptotic effect of BCL-2. Bax and Bak have been shown to play a critical role in cytochrome c release from mitochondria and thus initiate apoptosis. Bax exerts a pro-apoptotic rather than an anti-apoptotic effect on cells. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. In most follicular lymphomas, neoplastic germinal centers express high levels of BCL-2 alpha protein, whereas the normal or hyperplastic germinal centers are negative. Two transcript variants of BCL-2, produced by alternate splicing, differ in their C-terminal ends. The overexpression of BCL-2 has been linked to human cancers such as B-cell lymphoma and prostate cancer.Especificaciones
| Bcl2 | |
| Monoclonal | |
| 1.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P10417 | |
| Bcl2 | |
| Bcl2 Fusion Protein Ag24261 | |
| 150 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 1B5B9 | |
| Unconjugated | |
| Bcl2 | |
| AW986256, B cell leukemia/lymphoma 2, Bcl2, C430015F12Rik, D630044D05Rik, D830018M01Rik | |
| Mouse | |
| Protein G | |
| RUO | |
| 12043 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido