missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
This gene is a member of the Bardet-Biedl syndrome (BBS) gene family. Bardet-Biedl syndrome is an autosomal recessive disorder characterized by severe pigmentary retinopathy, obesity, polydactyly, renal malformation and mental retardation. The proteins encoded by BBS gene family members are structurally diverse and the similar phenotypes exhibited by mutations in BBS gene family members is likely due to their shared roles in cilia formation and function. Many BBS proteins localize to the basal bodies, ciliary axonemes, and pericentriolar regions of cells. BBS proteins may also be involved in intracellular trafficking via microtubule-related transport. The protein encoded by this gene forms a multiprotein BBSome complex with six other BBS proteins.
Especificaciones
Especificaciones
| Antígeno | BBS2 |
| Aplicaciones | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.33 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | BBS2 |
| N.º de referencia del gen | Q99MH9, Q9BXC9, Q9CWF6 |
| Alias de gen | Bardet Biedl syndrome 2, BBS, BBS2 |
| Símbolos de los genes | BBS2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?